2,4-Dinitrotoluene ≥97%

Danger

Synonyms: 1,3-Dinitro-4-methylbenzene

101397-5G 101397-1KG 101397-100G
220032-099EA 43.08 USD
220032-099 220032-098 220032-097
2,4-Dinitrotoluene ≥97%
2,4-Dinitrotoluene
Formula: C₇H₆N₂O₄
MW: 182.14 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 67…70 °C
Density: 1.379 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007172
CAS Number: 121-14-2
EINECS: 204-450-0
UN: 3454
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR