Benzothiazole

Synonyms: Benzo[d]thiazole

101338-5G 101338-100G 101338-500G
220033-190EA 37.69 USD
220033-190 220033-188 220033-189
Benzothiazole
Benzothiazole
Formula: C₇H₅NS
MW: 135.19 g/mol
Boiling Pt: 231 °C (1013 hPa)
Density: 1.246 g/cm³ (20 °C)
MDL Number: MFCD00005775
CAS Number: 95-16-9
EINECS: 202-396-2
UN: 2810
ADR: 6.1,III
Merck Index: 13,01108

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR