Triphenylmethane ≥99%

101303-100G 101303-25G
220033-186EA 95.09 USD
220033-186 220033-187
Triphenylmethane ≥99%
Triphenylmethane
Formula: C₁₉H₁₆
MW: 244.34 g/mol
Boiling Pt: 359 °C (1013 hPa)
Melting Pt: 90…92 °C
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004763
CAS Number: 519-73-3
EINECS: 208-275-0
Merck Index: 12,09871

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR