ω-Bromo-4-chloroacetophenone ≥98%

Synonyms: 2-Bromo-4'-chloroacetophenone, 2-Bromo-1-(4-chlorophenyl)ethanone, Bromomethyl p-chlorophenyl ketone, α-Bromo-4-chloroacetophenone, 2-Bromo-4′-chloroacetophenone, 4-Chlorophenacyl bromide

101273-100G 101273-25G
220033-183EA 187.38 USD
220033-183 220033-184
ω-Bromo-4-chloroacetophenone ≥98%
ω-Bromo-4-chloroacetophenone
Formula: C₈H₆BrClO
MW: 233.49 g/mol
Boiling Pt: 278 °C (1013 hPa)
Melting Pt: 110 °C
MDL Number: MFCD00000625
CAS Number: 536-38-9
EINECS: 208-631-5
UN: 3261
ADR: 8,III
Merck Index: 13,02172

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR