Pentafluorobenzyl bromide ≥99%

Warning

Synonyms: α-Bromo-2,3,4,5,6-pentafluorotoluene, 2,3,4,5,6-Pentafluorobenzyl bromide

101052-5G 101052-25G 101052-1G
220033-170EA 62.89 USD
220033-170 220033-169 220033-168
Pentafluorobenzyl bromide ≥99%
Pentafluorobenzyl bromide
Formula: (C₆F₅)CH₂Br
MW: 260.99 g/mol
Boiling Pt: 174 °C (1013 hPa)
Density: 1.72 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000299
CAS Number: 1765-40-8
EINECS: 217-182-4
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR