1-(2-Pyridylazo)-2-naphthol (PAN) indicator

Synonyms: PAN, 1-(2-Pyridylazo)-2-naphthol

101036-5G 101036-25G
220033-167EA 240.8 USD
220033-167 220033-166
1-(2-Pyridylazo)-2-naphthol (PAN) indicator
1-(2-Pyridylazo)-2-naphthol
Formula: C₁₅H₁₁N₃O
MW: 249.27 g/mol
Boiling Pt: 412 °C (1013 hPa)
Melting Pt: 137…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00004071
CAS Number: 85-85-8
EINECS: 201-637-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR