4-Bromoaniline ≥97%

Danger

100900-100G 100900-5G 100900-500G
220033-161EA 93.8 USD
220033-161 220033-163 220033-162
4-Bromoaniline ≥97%
4-Bromoaniline
Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 223 °C (1013 hPa)
Melting Pt: 62…64 °C
Density: 1.799 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007822
CAS Number: 106-40-1
EINECS: 203-393-9
UN: 2811
ADR: 6.1,III
Merck Index: 13,01388

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR