ω-Bromo-2-methoxyacetophenone ≥98%

Synonyms: 2-Methoxyphenacyl bromide, α-Bromo-2-methoxyacetophenone, 2-Bromo-2'-methoxyacetophenone, 2-Bromo-1-(2-methoxyphenyl)ethanone, 2-Bromo-1-(2-methoxyphenyl)ethan-1-one, Bromomethyl o-methoxyphenyl ketone

100854-5G 100854-10G 100854-1G
220033-156EA 242.31 USD
220033-156 220033-154 220033-155
ω-Bromo-2-methoxyacetophenone ≥98%
ω-Bromo-2-methoxyacetophenone
Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Boiling Pt: 324 °C (1013 hPa)
Melting Pt: 40…44 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000196
CAS Number: 31949-21-0
EINECS: 250-870-2
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR