4-Aminobenzoic acid ≥99%

Synonyms: Vitamin Bx, para-Aminobenzoic acid, PABA, Vitamin H1

100536-50G 100536-1KG 100536-250G
220033-142EA 33.71 USD
220033-142 220033-140 220033-141
4-Aminobenzoic acid ≥99%
4-Aminobenzoic acid
Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 340 °C (1013 hPa)
Melting Pt: 188 °C
Density: 1.45 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00007894
CAS Number: 150-13-0
EINECS: 205-753-0
Merck Index: 13,00422

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR