2,6-Dichlorobenzyl alcohol ≥99%

Synonyms: (2,6-Dichlorophenyl)methanol

100293-25G 100293-10G
220033-129EA 60.42 USD
220033-129 220033-128
2,6-Dichlorobenzyl alcohol ≥99%
2,6-Dichlorobenzyl alcohol
Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 96 - 98 °C
MDL Number: MFCD00004609
CAS Number: 15258-73-8
EINECS: 239-300-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR