3-Aminophenol ≥98%

Warning

Synonyms: 3-Hydroxyaniline, m-Aminophenol

100242-5G 100242-500G 100242-100G
220031-959EA 37.69 USD
220031-959 220031-958 220031-957
3-Aminophenol ≥98%
3-Aminophenol
Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 299 °C (1013 hPa)
Melting Pt: 122 °C
Density: 1.275 g/cm³ (20 °C)
Flash Pt: 92 °C
MDL Number: MFCD00007786
CAS Number: 591-27-5
EINECS: 209-711-2
UN: 2512
ADR: 6.1,III
Merck Index: 13,00459

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR