3-Iodobenzylamine ≥96%

Synonyms: (3-Iodophenyl)methanamine

100064-1G 100064-5G
220031-949EA 116.31 USD
220031-949 220031-950
3-Iodobenzylamine ≥96%
3-Iodobenzylamine
Formula: C₇H₈IN
MW: 233.05 g/mol
Boiling Pt: 132 °C (8 mmHg)
Density: 1.748 g/cm³ (20 °C)
MDL Number: MFCD00192227
CAS Number: 696-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR