(S)-(+)-Mandelic acid 99.0-100.5%

Danger

Synonyms: L(+)-Mandelic acid, (S)-(+)-2-Hydroxy-2-phenylacetic acid, L(+)-2-Hydroxy-2-phenylacetic acid, (S)-2-Hydroxy-2-phenylacetic acid

M2004-100G M2004-1KG M2004-25G-A M2004-500G
220197-225EA 49.32 USD
220197-225 220197-227 220197-228 220197-229
(S)-(+)-Mandelic acid 99.0-100.5%
(S)-(+)-Mandelic acid
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 130…132 °C
Density: 1.349 g/cm³ (20 °C)
Flash Pt: >190 °C
MDL Number: MFCD00004495
CAS Number: 17199-29-0
EINECS: 241-240-8

Order Now

Specification Test Results

Assay 99.0-100.5% (titration)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR