2'-Hydroxyacetanilide ≥97%

Synonyms: N-Acetyl-2-aminophenol, N-(2-Hydroxyphenyl)acetamide, 2-Acetamidophenol, Acetylcholine iodide

A7000-100G-A A7000-5G A7000-25G A7000-25G-A
220104-340EA 93.26 USD
220104-340 220104-343 220104-341 220104-342
2'-Hydroxyacetanilide ≥97%
2'-Hydroxyacetanilide
Formula: C₈H₉NO₂
MW: 151.16 g/mol
MDL Number: MFCD00002181
CAS Number: 614-80-2
EINECS: 210-396-9

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR