Zinc acetate dihydrate ≥98%, Reagent Grade

Warning

Z0625-1KG Z0625-500G
220240-206EA 62.77 USD
220240-206 220240-207
Zinc acetate dihydrate ≥98%, Reagent Grade
Zinc acetate dihydrate
Formula: (H₃CCOO)₂Zn·2H₂O
MW: 219.51 g/mol
Boiling Pt: 908 °C (1013 hPa)
Density: 1.735 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066961
CAS Number: 5970-45-6
EINECS: 209-170-2
UN: 3077
ADR: 9,III
Merck Index: 12,10256

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR