TRITON™ X-114 (Polyethylene glycol tert-octylphenyl ether), Lab-Grade

Danger

X114-100ML X114-1GA X114-500ML X114-5GA X114-1L
220239-688EA 48.69 USD
220239-688 220239-689 220240-390 220240-391 220239-690
TRITON™ X-114 (Polyethylene glycol tert-octylphenyl ether), Lab-Grade
TRITON™ X-114 (Polyethylene glycol tert-octylphenyl ether)
Boiling Pt: ∼150 °C (1013 hPa)
Density: 1.055 g/cm³ (20 °C)
Flash Pt: >93 °C
Storage Temperature: Ambient
MDL Number: MFCD00128254
CAS Number: 9036-19-5
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR