TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)

Danger

Synonyms: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol

ampouled under argon

X100PC-10X5ML X100PC-5ML X100PC-5X5ML
220240-024EA 1223.15 USD
220240-024 220239-670 220239-671
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
Boiling Pt: ? 200 °C (1013 hPa)
Melting Pt: 6 °C
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 251 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00132505
CAS Number: 9002-93-1
UN: 3082
ADR: 9,III
Merck Index: 13,06793

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR