Pyruvic acid (natural) ≥80% in aqueous solution

Synonyms: α-Ketopropionic acid, 2-Oxopropionic acid, 2-Ketopropionic acid, 2-Oxopropanoic acid

W297070-1KG-K W297070-100G-K W297070-25G-K
220233-289EA 2323.08 USD
220233-289 220233-288 220233-290
Pyruvic acid (natural) ≥80% in aqueous solution
Pyruvic acid
Formula: C₃H₄O₃
MW: 88.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00002585
CAS Number: 127-17-3
EINECS: 204-824-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR