Pyruvic acid ≥97% Kosher

Danger

Synonyms: α-Ketopropionic acid, 2-Oxopropionic acid, 2-Ketopropionic acid, 2-Oxopropanoic acid

W297003-1KG-K W297003-5KG-K W297003-250G-K W297003-25KG-K
220233-283EA 250.77 USD
220233-283 220233-286 220233-284 220233-285
Pyruvic acid ≥97% Kosher
Pyruvic acid
Formula: C₃H₄O₃
MW: 88.06 g/mol
Boiling Pt: 165 °C (1013 hPa)
Melting Pt: 12 °C
Density: 1.58 g/cm³ (20 °C)
Flash Pt: 82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002585
CAS Number: 127-17-3
EINECS: 204-824-3
UN: 3265
ADR: 8,II
Merck Index: 12,08205

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR