Methyl-N-methylanthranilate (natural) ≥98%

Warning

Synonyms: Methyl N-Methylanthranilate, Dimethyl anthranilate

W271810-1KG-K W271810-100G-K
220231-802EA 1615.38 USD
220231-802 220231-801
Methyl-N-methylanthranilate (natural) ≥98%
Methyl-N-methylanthranilate
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Density: 1.12 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00017183
CAS Number: 85-91-6
EINECS: 201-642-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR