Methyl-N-methylanthranilate ≥97%

Warning

Synonyms: Methyl N-Methylanthranilate, Dimethyl anthranilate

W271802-5KG-K W271802-100G-K W271802-1KG-K
220231-799EA 849.23 USD
220231-799 220231-797 220231-798
Methyl-N-methylanthranilate ≥97%
Methyl-N-methylanthranilate
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Density: 1.12 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00017183
CAS Number: 85-91-6
EINECS: 201-642-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR