Benzoic acid methyl ester (natural) ≥98%

Warning

Synonyms: Methyl benzoate

W268313-5KG-K W268313-1KG-K W268313-100G-K
220229-748EA 1240 USD
220229-748 220229-747 220229-746
Benzoic acid methyl ester (natural) ≥98%
Benzoic acid methyl ester
Formula: C₆H₅COOCH₃
MW: 136.15 g/mol
Boiling Pt: 199.6 °C (1013 hPa)
Melting Pt: -12 °C
Density: 1.0888 g/cm³ (20 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00008421
CAS Number: 93-58-3
EINECS: 202-259-7
Merck Index: 13,06052
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=287e16081e44f5525ff8b4d34f0a6fece44a3073&vid=2be9af3b90e55af477c374c091cd0888dcf60b96&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR