4-(4-Methoxyphenyl)-2-butanone ≥98% FCC

Danger

Synonyms: 1-(4-Methoxyphenyl)-3-butanone, Anisylacetone

W267201-100G-K W267201-1KG-K W267201-5KG-K
220232-379EA 90.92 USD
220232-379 220232-380 220232-381
4-(4-Methoxyphenyl)-2-butanone ≥98% FCC
4-(4-Methoxyphenyl)-2-butanone
Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 72 °C (0.02 mmHg)
Melting Pt: 10 °C
Density: 1.0619 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008791
CAS Number: 104-20-1
EINECS: 203-184-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR