p-Anisaldehyde ≥97.5% FCC

Synonyms: 4-Methoxybenzaldehyde

W267007-1KG-K W267007-25KG-K W267007-5KG-K
220232-366EA 110.77 USD
220232-366 220232-367 220232-368
p-Anisaldehyde ≥97.5% FCC
p-Anisaldehyde
Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: 0 - 2 °C
Density: 1.12 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003385
CAS Number: 123-11-5
EINECS: 204-602-6
Merck Index: 13,00666

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR