Geraniol (natural)

Warning

Synonyms: (E)-3,7-Dimethylocta-2,6-dien-1-ol

W250716-9KG-K W250716-4KG-K W250716-1KG-K
220231-564EA 2169.23 USD
220231-564 220231-563 220231-562
Geraniol (natural)
Geraniol
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
MDL Number: MFCD00002917
CAS Number: 106-24-1
EINECS: 203-377-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR