Ethyl laurate (natural) ≥98%

Synonyms: Dodecanoic acid ethyl ester, Ethyl dodecanoate, Ethyl Laurate, Dodecylic acid ethyl ester, Lauric acid ethyl ester

W244112-100G-K W244112-1KG-K W244112-4KG-K
220231-696EA 283.08 USD
220231-696 220231-697 220231-698
Ethyl laurate (natural) ≥98%
Ethyl laurate
Formula: C₁₄H₂₈O₂
MW: 228.38 g/mol
Boiling Pt: 269 °C (1013 hPa)
Melting Pt: –2 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00015065
CAS Number: 106-33-2
EINECS: 203-386-0
Merck Index: 13,03851

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR