Ethyl acetoacetate (natural) ≥99%

Warning

Synonyms: Acetoacetic acid ethyl ester, EAA

W241512-1KG-K W241512-100G-K W241512-5KG-K
220229-914EA 318.46 USD
220229-914 220229-913 220229-915
Ethyl acetoacetate (natural) ≥99%
Ethyl acetoacetate
Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 180.8 °C (1013 hPa)
Melting Pt: -45 °C
Density: 1.0188 g/cm³ (20 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00009199
CAS Number: 141-97-9
EINECS: 205-516-1
Merck Index: 13,03793

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR