(±)-Citronellal (natural) ≥85%

Synonyms: (+/-)-Citronellal, (±)-3,7-Dimethyloct-6-enal, (+/-)-3,7-Dimethyl-6-octenal

W230715-1KG W230715-8KG W230715-4KG
220230-661EA 80 USD
220230-661 220230-663 220230-662
(±)-Citronellal (natural) ≥85%
(±)-Citronellal
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 207 °C (1013 hPa)
Melting Pt: –15 °C
Density: 0.8548 g/cm³ (20 °C)
MDL Number: MFCD00038090
CAS Number: 106-23-0
EINECS: 203-376-6
Merck Index: 13,02353

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR