(±)-Citronellal ≥80% Kosher

Synonyms: (+/-)-Citronellal, (±)-3,7-Dimethyloct-6-enal, (+/-)-3,7-Dimethyl-6-octenal

W230707-1KG-K W230707-4KG-K W230707-8KG-K
220230-657EA 247.69 USD
220230-657 220230-658 220230-659
(±)-Citronellal ≥80% Kosher
(±)-Citronellal
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 207 °C (1013 hPa)
Melting Pt: –15 °C
Density: 0.8548 g/cm³ (20 °C)
MDL Number: MFCD00038090
CAS Number: 106-23-0
EINECS: 203-376-6
Merck Index: 13,02353

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR