Isobutyl butyrate (natural) ≥98%

Synonyms: Butyric acid Isobutyl ester, 2-Methyl-1-propylbutyrate

W218715-1KG-K W218715-100G-K W218715-4KG-K
220230-943EA 290.77 USD
220230-943 220230-942 220230-944
Isobutyl butyrate (natural) ≥98%
Isobutyl butyrate
Formula: C₈H₁₆O₂
MW: 144.21 g/mol
MDL Number: MFCD00027134
CAS Number: 539-90-2
EINECS: 208-729-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR