Isobutyl butyrate ≥98% Kosher

Synonyms: Butyric acid Isobutyl ester, 2-Methyl-1-propylbutyrate

W218707-9KG-K W218707-1KG-K
220230-940EA 512.31 USD
220230-940 220230-938
Isobutyl butyrate ≥98% Kosher
Isobutyl butyrate
Formula: C₈H₁₆O₂
MW: 144.21 g/mol
MDL Number: MFCD00027134
CAS Number: 539-90-2
EINECS: 208-729-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR