Isobutanol ≥99% Kosher

Danger

W217905-20KG-K W217905-2KG-K W217905-8KG-K
220230-842EA 533.85 USD
220230-842 220230-843 220230-844
Isobutanol ≥99% Kosher
Isobutanol
Formula: (CH₃)₂CHCH₂OH
Boiling Pt: 107.9 °C (1013 hPa)
Melting Pt: –108 °C
Density: 0.802 g/cm³ (20 °C)
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00004740
CAS Number: 78-83-1
EINECS: 201-148-0
UN: 1212
ADR: 3,III
Merck Index: 13,05148
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de7c83a107c7a71fa54ed2b1bbdfb78617fe3f67&vid=df3ce90fc7ae8dafb7fabe31b244825735325030&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR