Benzyl benzoate ≥99%, halal

Warning

Synonyms: Benzoic acid benzyl ester

W213802-10KG-K W213802-1KG-K W213802-5KG-K
220230-621EA 329.23 USD
220230-621 220230-622 220230-623
Benzyl benzoate ≥99%, halal
Benzyl benzoate
Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: 17…20 °C
Density: 1.118 g/cm³ (20 °C)
Flash Pt: 158 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003075
CAS Number: 120-51-4
EINECS: 204-402-9
UN: 3082
ADR: 9,III
Merck Index: 13,01129

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR