trans-Anethole ≥99% FCC

Synonyms: (E)-1-Methoxy-4-(prop-1-en-1-yl)benzene

W208604-10KG-K W208604-1KG-K W208604-5KG-K
220230-495EA 1009.23 USD
220230-495 220230-496 220230-497
trans-Anethole ≥99% FCC
trans-Anethole
Formula: C₁₀H₁₂O
MW: 148.2 g/mol
Boiling Pt: 236 °C (1013 hPa)
Melting Pt: 21…23 °C
Density: 0.9883 g/cm³ (20 °C)
Flash Pt: 96 °C
MDL Number: MFCD00009284
CAS Number: 4180-23-8
EINECS: 224-052-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR