Isopentyl benzoate ≥98% FCC

Synonyms: Isoamyl Benzoate, Benzoic acid isopentyl ester

W205818-10KG-K W205818-1KG-K
220230-409EA 876.92 USD
220230-409 220230-410
Isopentyl benzoate ≥98% FCC
Isopentyl benzoate
Formula: C₁₂H₁₆O₂
MW: 192.26 g/mol
Density: 0.99 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00026515
CAS Number: 94-46-2
EINECS: 202-334-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR