L(+)-Valine ≥98%, Reagent Grade

V0500-100G V0500-1KG V0500-1G V0500-25G V0500-500G
220229-434EA 118.52 USD
220229-434 220229-436 220229-435 220229-437 220229-438
L(+)-Valine ≥98%, Reagent Grade
L(+)-Valine
Formula: C₅H₁₁NO₂
Boiling Pt: 217 °C (1013 hPa)
Melting Pt: 86 °C
Density: 1.284 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064220
CAS Number: 72-18-4
EINECS: 200-773-6
Merck Index: 13,09458

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR