1-Undecanol ≥99%

Synonyms: 1-Undecyl alcohol, Undecyl alcohol, Undecan-1-ol, Decyl carbinol, 1-Hydroxy undecane

U1001-100G U1001-500G
220228-652EA 63.54 USD
220228-652 220228-653
1-Undecanol ≥99%
1-Undecanol
Formula: C₁₁H₂₄O
MW: 172.31 g/mol
Boiling Pt: 243 °C (1013 hPa)
Melting Pt: 13 - 15 °C
Density: 0.83 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00004751
CAS Number: 112-42-5
EINECS: 203-970-5
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR