TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) for molecular biology

Danger

Synonyms: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol

T8787-100ML T8787-250ML T8787-50ML
220228-183EA 65.57 USD
220228-183 220228-184 220228-185
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) for molecular biology
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
Boiling Pt: ? 200 °C (1013 hPa)
Melting Pt: 6 °C
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 251 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00132505
CAS Number: 9002-93-1
UN: 3082
ADR: 9,III
Merck Index: 13,06793

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR