1,1,1-Tris(hydroxymethyl)ethane ≥99%

Synonyms: 2-(Hydroxymethyl)-2-methyl-1,3-propanediol, Ethylidynetrimethanol

T87807-2KG T87807-500G T87807-12KG
220228-173EA 146.46 USD
220228-173 220228-174 220228-172
1,1,1-Tris(hydroxymethyl)ethane ≥99%
1,1,1-Tris(hydroxymethyl)ethane
Formula: C₅H₁₂O₃
MW: 120.15 g/mol
Boiling Pt: 287 °C (1013 hPa)
MDL Number: MFCD00004687
CAS Number: 77-85-0
EINECS: 201-063-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR