L-Threonine ≥98%, Reagent Grade

Synonyms: (2S,3R)-(-)-Threonine, H-Thr-OH, (2S,3R)-2-Amino-3-hydroxybutyric acid

T8625-5KG T8625-100G T8625-1KG T8625-25G T8625-500G T8625-1G T8625-10G
220228-132EA 7224.62 USD
220228-132 220228-126 220228-129 220228-130 220228-131 220228-128 220228-127
L-Threonine ≥98%, Reagent Grade
L-Threonine
Formula: C₄H₉NO₃
MW: 119.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064270
CAS Number: 72-19-5
EINECS: 200-774-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR