Triphenyl phosphite ≥97%

Warning

T84654-5G T84654-500G T84654-3KG
220228-089EA 20.08 USD
220228-089 220228-088 220228-087
Triphenyl phosphite ≥97%
Triphenyl phosphite
Formula: C₁₈H₁₅O₃P
MW: 310.29 g/mol
Boiling Pt: 360 °C (1013 hPa)
Melting Pt: 22…25 °C
Density: 1.19 g/cm³ (20 °C)
Flash Pt: 188 °C
Storage Temperature: Ambient
MDL Number: MFCD00003032
CAS Number: 101-02-0
EINECS: 202-908-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR