Triphenylphosphine ≥99%, ReagentPlus®

Warning

T84409-25G T84409-1KG T84409-500G T84409-100G T84409-1G
220228-072EA 26.15 USD
220228-072 220228-071 220228-073 220228-069 220228-070
Triphenylphosphine ≥99%, ReagentPlus®
Triphenylphosphine
Formula: C₁₈H₁₅P
MW: 262.29 g/mol
Boiling Pt: 377 °C (1013 hPa)
Melting Pt: 79 °C
Density: 1.194 g/cm³ (20 °C)
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00003043
CAS Number: 603-35-0
EINECS: 210-036-0
Merck Index: 12,09873

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR