Trityl chloride ≥98%

Danger

Synonyms: Trt-Cl, Chlorotriphenylmethane, Triphenylmethyl chloride, (Chloromethanetriyl)tribenzene, Triphenylchloromethane

T83801-100G T83801-500G T83801-25G
220228-054EA 95.31 USD
220228-054 220228-056 220228-055
Trityl chloride ≥98%
Trityl chloride
Formula: C₁₉H₁₅Cl
MW: 278.78 g/mol
Boiling Pt: 374 °C (1013 hPa)
Melting Pt: 109…112 °C
Density: 1.25 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000813
CAS Number: 76-83-5
EINECS: 200-986-4
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR