Thiourea ≥99.0%, ReagentPlus®

Warning

T7875-1KG T7875-5G T7875-5KG T7875-100G T7875-500G
220227-973EA 74.38 USD
220227-973 220227-975 220227-976 220227-972 220227-974
Thiourea ≥99.0%, ReagentPlus®
Thiourea
Formula: NH₂CSNH₂
Melting Pt: 174…177 °C
Density: 1.435 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008067
CAS Number: 62-56-6
EINECS: 200-543-5
UN: 3077
ADR: 9,III
Merck Index: 13,09443
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fff6222c54fe16ea4d36edcbf320745cb1e18d6&vid=e9ae5dc49e92d68f893244a5cd4e0245f289ed09&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR