Trimethylammonium chloride

Warning

Synonyms: Trimethylamine hydrochloride

T72761-100G T72761-25G T72761-500G
220227-827EA 33.17 USD
220227-827 220227-828 220227-829
Trimethylammonium chloride
Trimethylammonium chloride
Formula: C₃H₁₀ClN
MW: 95.57 g/mol
Melting Pt: 273 - 278 °C
Density: ~ 1 g/cm³ (20 °C)
MDL Number: MFCD00012478
CAS Number: 593-81-7
EINECS: 209-810-0
Merck Index: 12,09841

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR