Pivalic acid ≥99%

Synonyms: 2,2-Dimethylpropionic acid, Trimethylacetic acid

T71803-100ML T71803-5ML T71803-500ML
220227-803EA 40.06 USD
220227-803 220227-805 220227-804
Pivalic acid ≥99%
Pivalic acid
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 164 °C (1013 hPa)
Melting Pt: 32 - 35 °C
Density: 0.91 g/cm³ (20 °C)
Flash Pt: 640 °C
Storage Temperature: Ambient
MDL Number: MFCD00004194
CAS Number: 75-98-9
EINECS: 200-922-5
UN: 3261
ADR: 8,III
Merck Index: 12,07667

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR