2,2,2-Trifluoroethanol ≥99%, ReagentPlus®

Danger

Synonyms: Trifluoro Ethanol, TFE

T63002-100G T63002-25G
220227-529EA 19.32 USD
220227-529 220227-531 220227-530
2,2,2-Trifluoroethanol ≥99%, ReagentPlus®
2,2,2-Trifluoroethanol
Formula: C₂H₃F₃O
MW: 100.04 g/mol
Boiling Pt: 74.1 °C (1013 hPa)
Melting Pt: –43.5 °C
Density: 1.39 g/cm³ (20 °C)
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00004672
CAS Number: 75-89-8
EINECS: 200-913-6
UN: 2929
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR