Triethyl orthoformate ≥98%, Reagent Grade

Danger

T60453-500ML T60453-1L
220227-439EA 23.66 USD
220227-439 220227-437
Triethyl orthoformate ≥98%, Reagent Grade
Triethyl orthoformate
Formula: C₇H₁₆O₃
MW: 148.2 g/mol
Boiling Pt: 146 °C (1013 hPa)
Melting Pt: –61 °C
Density: 0.8964 g/cm³ (20 °C)
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00009230
CAS Number: 122-51-0
EINECS: 204-550-4
UN: 2524
ADR: 3,III
Merck Index: 12,07012

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR