1,2,4-Triazole

Warning

Synonyms: 1,2,4-1H-Triazole, 1H-1,2,4-Triazole

T46108-100G T46108-500G T46108-25G
220222-787EA 101.23 USD
220222-787 220222-789 220222-788
1,2,4-Triazole
1,2,4-Triazole
Formula: C₂H₃N₃
MW: 69.07 g/mol
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 116…119 °C
Density: 1.394 g/cm³ (25 °C)
MDL Number: MFCD00005228
CAS Number: 288-88-0
EINECS: 206-022-9
Merck Index: 12,09735

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR