p-Toluenesulfonyl chloride ≥98%, Reagent Grade

Danger

Synonyms: Toluene-4-sulphonyl chloride, Tosyl chloride, 4-Toluenesulphonyl chloride, Toluene-p-sulphonyl chloride

T35955-1KG T35955-3KG
220226-747EA 78.46 USD
220226-747 220226-748
p-Toluenesulfonyl chloride ≥98%, Reagent Grade
p-Toluenesulphonyl chloride
Formula: C₇H₇ClO₂S
MW: 190.65 g/mol
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: 67 °C
Density: 1.49 g/cm³ (20 °C)
Flash Pt: 128 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00007450
CAS Number: 98-59-9
EINECS: 202-684-8
UN: 3261
ADR: 8,II
Merck Index: 13,09612

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR